5-Brombenzothiazol 98%

Lieferant: Apollo Scientific
Achtung

OR015035-1G OR015035-25G OR015035-5G
APOSOR015035-1GEA 36.4 EUR
APOSOR015035-1G APOSOR015035-25G APOSOR015035-5G
5-Brombenzothiazol 98%
5-Bromobenzothiazole
Formel: C₇H₄BrNS
Molecular Weight: 214.09 g/mol
Schmelzpunkt: 105…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03001442
CAS-Nummer: 768-11-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR