1-Phenyl-2-(pyrazin-2-yl)ethanol 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR0156-1G
OR0156-5G
OR0156-10G
APOSOR0156-1GEA
163
EUR
APOSOR0156-1G
APOSOR0156-5G
APOSOR0156-10G
1-Phenyl-2-(pyrazin-2-yl)ethanol 95+%
1-Phenyl-2-(pyrazin-2-yl)ethanol
Formel:
C₁₂H₁₂N₂O Molecular Weight: 200,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00235147 CAS-Nummer: 36914-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...