1-Pyridin-3-ylethylamine

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-(1-Aminoethyl)pyridine, 1-(Pyridin-3-yl)ethanamine

OR01562-1G OR01562-250MG
APOSOR01562-1GEA 33.5 EUR
APOSOR01562-1G APOSOR01562-250MG
1-Pyridin-3-ylethylamine
1-Pyridin-3-ylethylamine
Formel: C₇H₁₀N₂
Molecular Weight: 122,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01691641
CAS-Nummer: 56129-55-6
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR