2',5'-Dihydroxyacetophenon 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(2,5-Dihydroxyphenyl)ethan-1-on

OR01582-100G OR01582-25G OR01582-500G
APOSOR01582-100GEA 43.5 EUR
APOSOR01582-100G APOSOR01582-25G APOSOR01582-500G
2',5'-Dihydroxyacetophenon 98%
2',5'-Dihydroxyacetophenon
Formel: C₈H₈O₃
Molecular Weight: 152,15 g/mol
Schmelzpunkt: 204…208 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002343
CAS-Nummer: 490-78-8
EINECS: 207-716-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR