1-Phenyl-2-(quinoxalin-2-yl)ethanol 95+%

Lieferant: Apollo Scientific

OR0160-10G OR0160-1G OR0160-5G
APOSOR0160-10GEA 606 EUR
APOSOR0160-10G APOSOR0160-1G APOSOR0160-5G
1-Phenyl-2-(quinoxalin-2-yl)ethanol 95+%
1-Phenyl-2-(quinoxalin-2-yl)ethanol
Formel: C₁₆H₁₄N₂O
Molecular Weight: 250,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03426052
CAS-Nummer: 849021-27-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR