Oxalic acid mono(N-methyl)amide 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-Methyloxamic acid, 2-(Methylamino)-2-oxoacetic acid
OR016302-5G
OR016302-1G
APOSOR016302-5GEA
132
EUR
APOSOR016302-5G
APOSOR016302-1G
Oxalic acid mono(N-methyl)amide 95+%
Oxalic acid mono(N-methyl)amide
Formel:
C₃H₅NO₃ Molecular Weight: 103,08 g/mol Schmelzpunkt: 144…150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06245498 CAS-Nummer: 29262-58-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...