3-(Aminomethyl)-2-tert-butoxypyridine 95+%

Lieferant: Apollo Scientific
Achtung

OR01639-1G OR01639-5G
APOSOR01639-1GEA 265 EUR
APOSOR01639-1G APOSOR01639-5G
3-(Aminomethyl)-2-tert-butoxypyridine 95+%
3-(Aminomethyl)-2-tert-butoxypyridine
Formel: C₁₀H₁₆N₂O
Molecular Weight: 180.25 g/mol
Siedepunkt: 52…54 °C/0.37mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD06245525
CAS-Nummer: 849021-22-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR