3-Amino-3-methylbutanoic acid

Lieferant: Apollo Scientific
Achtung

OR01641-25G OR01641-5G OR01641-1G
APOSOR01641-25GEA 102 EUR
APOSOR01641-25G APOSOR01641-5G APOSOR01641-1G
3-Amino-3-methylbutanoic acid
3-Amino-3-methylbutanoic acid
Formel: C5H11NO2
Molecular Weight: 117.1500 g/mol
Schmelzpunkt: 232…233 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06245495
CAS-Nummer: 625-05-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR