1-N-Boc-4-(2-furfurylmethylamino)piperidine 95+%
Lieferant: Apollo Scientific
Synonyme:
4-[(Fur-2-ylmethyl)amino]piperidine, N1-BOC protected
OR01644-1G
OR01644-5G
OR01644-25G
APOSOR01644-1GEA
229
EUR
APOSOR01644-1G
APOSOR01644-5G
APOSOR01644-25G
1-N-Boc-4-(2-furfurylmethylamino)piperidine 95+%
1-N-Boc-4-(2-furfurylmethylamino)piperidine
Formel:
C₁₅H₂₄N₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06245534 CAS-Nummer: 883516-51-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...