3-Chlorbenzophenon
Lieferant: Apollo Scientific
Synonyme:
m-Chlorobenzophenone
OR016460-100G
APOSOR016460-100GEA
126
EUR
APOSOR016460-100G
3-Chlorbenzophenon
3-Chlorbenzophenon
Formel:
C₁₃H₉ClO Molecular Weight: 216,67 g/mol Schmelzpunkt: 81…83 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009816 CAS-Nummer: 1016-78-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...