2-[Benzyl(2-hydroxyethyl)amino]-1-ethanol 95+%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2,2-(Benzylimino)diethanol

OR01671-10G OR01671-1G OR01671-5G
APOSOR01671-10GEA 139 EUR
APOSOR01671-10G APOSOR01671-1G APOSOR01671-5G
2-[Benzyl(2-hydroxyethyl)amino]-1-ethanol 95+%
2-[Benzyl(2-hydroxyethyl)amino]-1-ethanol
Formel: C₁₁H₁₇NO₂
Molecular Weight: 195,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00020587
CAS-Nummer: 101-32-6
EINECS: 202-934-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR