N2-[(Benzyloxy)carbonyl]-L-asparagin 97%
Lieferant: Apollo Scientific
Synonyme:
Z-Asn-OH, Nalpha-Cbz-L-asparagine
OR01678-100G
OR01678-25G
OR01678-500G
APOSOR01678-100GEA
63.8
EUR
APOSOR01678-100G
APOSOR01678-25G
APOSOR01678-500G
N2-[(Benzyloxy)carbonyl]-L-asparagin 97%
N-Cbz-L-asparagine
Formel:
C₁₂H₁₄N₂O₅ Molecular Weight: 266,3 g/mol Schmelzpunkt: 160…164 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008035 CAS-Nummer: 2304-96-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...