3-Aminobenzolsulfonamid

Lieferant: Apollo Scientific
Achtung

Synonyme: Anilin-3-sulfonsäureamid, 3-Aminobenzolsulfonsäureamid

OR017076-100G OR017076-25G
APOSOR017076-100GEA 140 EUR
APOSOR017076-100G APOSOR017076-25G
3-Aminobenzolsulfonamid
3-Aminobenzolsulfonamid
Formel: C₆H₈N₂O₂S
Molecular Weight: 172,21 g/mol
Schmelzpunkt: 139…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00035781
CAS-Nummer: 98-18-0
EINECS: 202-646-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR