2,3,4,5-Tetrahydro-1H-benzo[e][1,4]diazepine 97%

Lieferant: Apollo Scientific
Gefahr

OR01713-1G OR01713-5G
APOSOR01713-1GEA 0 EUR
APOSOR01713-1G APOSOR01713-5G
2,3,4,5-Tetrahydro-1H-benzo[e][1,4]diazepine 97%
2,3,4,5-Tetrahydro-1H-benzo[e][1,4]diazepine
Formel: C₉H₁₂N₂
Molecular Weight: 148,21 g/mol
Schmelzpunkt: 96…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03789577
CAS-Nummer: 5946-39-4
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR