1-Phenyl-2-imidazolidinon

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Phenylimidazolidin-2-one

OR01734-1G OR01734-250MG OR01734-5G
APOSOR01734-1GEA 434 EUR
APOSOR01734-1G APOSOR01734-250MG APOSOR01734-5G
1-Phenyl-2-imidazolidinon
1-Phenyl-2-imidazolidinon
Formel: C₉H₁₀N₂O
Molecular Weight: 162,1885 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02671066
CAS-Nummer: 1848-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR