3-Methylcinnolin-5-ylamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Amino-3-methylcinnoline

OR01737-1G OR01737-250MG OR01737-5G
APOSOR01737-1GEA 212 EUR
APOSOR01737-1G APOSOR01737-250MG APOSOR01737-5G
3-Methylcinnolin-5-ylamine
3-Methylcinnolin-5-ylamine
Formel: C₉H₉N₃
Molecular Weight: 159,19 g/mol
Schmelzpunkt: 189…191 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00466524
CAS-Nummer: 300690-74-8
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR