2-(3,5-Dimethyl-1H-pyrazol-1-yl)benzoesäure

Lieferant: Apollo Scientific
Achtung

OR01748-1G OR01748-500MG OR01748-50MG
APOSOR01748-1GEA 221 EUR
APOSOR01748-1G APOSOR01748-500MG APOSOR01748-50MG
2-(3,5-Dimethyl-1H-pyrazol-1-yl)benzoesäure
2-(3,5-Dimethyl-1H-pyrazol-1-yl)benzoic acid
Formel: C₁₂H₁₂N₂O₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 27363-77-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR