5-Amino-1-methyl-1H-pyrazole-4-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR01763-25G
APOSOR01763-25GEA
66.7
EUR
APOSOR01763-25G
5-Amino-1-methyl-1H-pyrazole-4-carbonitrile
5-Amino-1-methyl-1H-pyrazole-4-carbonitrile
Formel:
C₅H₆N₄ Molecular Weight: 122.13 g/mol Schmelzpunkt: 221…223 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00114569 CAS-Nummer: 5334-41-8 EINECS: 610-989-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...