4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-1-methyl-1H-pyrazole-3-carboxaldehyde

OR017852-5G OR017852-1G
APOSOR017852-5GEA 177 EUR
APOSOR017852-5G APOSOR017852-1G
4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde 95%
4-Bromo-1-methyl-1H-pyrazole-3-carbaldehyde
Formel: C₅H₅BrN₂O
Molecular Weight: 189,02 g/mol
Schmelzpunkt: 69…76 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00103235
CAS-Nummer: 287917-96-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR