3-tert-Butoxy carbonylaminophenyl acetic acid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Boc-3-Aminophenylacetic acid

OR0181-1G OR0181-5G OR0181-25G
APOSOR0181-1GEA 168 EUR
APOSOR0181-1G APOSOR0181-5G APOSOR0181-25G
3-tert-Butoxy carbonylaminophenyl acetic acid 95+%
3-tert-Butoxy carbonylaminophenyl acetic acid
Formel: C₁₃H₁₇NO₄
Molecular Weight: 251,28 g/mol
Schmelzpunkt: 100…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02682313
CAS-Nummer: 123036-51-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR