1-Oxiran-2-ylethanone

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Acetyloxirane

OR01831-1G OR01831-250MG OR01831-500MG
APOSOR01831-1GEA 432 EUR
APOSOR01831-1G APOSOR01831-250MG APOSOR01831-500MG
1-Oxiran-2-ylethanone
1-Oxiran-2-ylethanone
Formel: C₄H₆O₂
Molecular Weight: 86,09 g/mol
Siedepunkt: 45…46 °C/30mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00829205
CAS-Nummer: 4401-11-0
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR