Thieno[3,2-b]thiophene-2-carboxylic acid 95+%

Lieferant: Apollo Scientific
Achtung

OR0190-1G OR0190-5G
APOSOR0190-1GEA 65.4 EUR
APOSOR0190-1G APOSOR0190-5G
Thieno[3,2-b]thiophene-2-carboxylic acid 95+%
Thieno[3,2-b]thiophene-2-carboxylic acid
Formel: C₇H₄O₂S₂
Molecular Weight: 184.23 g/mol
Schmelzpunkt: 225…227 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00115174
CAS-Nummer: 1723-27-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR