3-(Methylthio)-1H-1,2,4-triazol

Lieferant: Apollo Scientific
Achtung

OR019167-1G OR019167-5G
APOSOR019167-1GEA 102 EUR
APOSOR019167-1G APOSOR019167-5G
3-(Methylthio)-1H-1,2,4-triazol
3-(Methylthio)-1H-1,2,4-triazol
Formel: C₃H₅N₃S
Schmelzpunkt: 98…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01312671
CAS-Nummer: 7411-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR