Methyl-4-(dimethoxymethyl)benzoate 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-formylbenzoate dimethyl acetal

OR0192-1G OR0192-5G OR0192-25G
APOSOR0192-1GEA 195 EUR
APOSOR0192-1G APOSOR0192-5G APOSOR0192-25G
Methyl-4-(dimethoxymethyl)benzoate 95+%
Methyl-4-(dimethoxymethyl)benzoate
Formel: C₁₁H₁₄O₄
Molecular Weight: 210,229 g/mol
Siedepunkt: 144 °C/2,25mmHg
Lagertemperatur: Raumtemperatur
CAS-Nummer: 42228-16-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR