1-(2,6-Dichlorphenyl)biguanid Hydrochlorid

Lieferant: Apollo Scientific
Achtung

OR01940-100G OR01940-25G OR01940-5G
APOSOR01940-100GEA 1080 EUR
APOSOR01940-100G APOSOR01940-25G APOSOR01940-5G
1-(2,6-Dichlorphenyl)biguanid Hydrochlorid
1-(2,6-Dichlorphenyl)biguanid Hydrochlorid
Formel: C₈H₁₀Cl₃N₅
Schmelzpunkt: 228…231 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 42823-15-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR