N-Methylindole-6-carboxylic acid methyl ester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 1-methyl-1H-indole-6-carboxylate, Methyl 1-Methylindole-6-carboxylate

OR0195-1G OR0195-5G
APOSOR0195-1GEA 21.9 EUR
APOSOR0195-1G APOSOR0195-5G
N-Methylindole-6-carboxylic acid methyl ester 98%
N-Methylindole-6-carboxylic acid methyl ester
Formel: C₁₁H₁₁NO₂
Molecular Weight: 189,22 g/mol
Schmelzpunkt: 90…93 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06245515
CAS-Nummer: 1204-32-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR