6-Hydroxy-m-anisidin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-4-methoxyphenol
OR01962-1G
OR01962-5G
APOSOR01962-1GEA
21.9
EUR
APOSOR01962-1G
APOSOR01962-5G
6-Hydroxy-m-anisidin
6-Hydroxy-m-anisidin
Formel:
C₇H₉NO₂ Molecular Weight: 139,16 g/mol Schmelzpunkt: 135…140 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06616911 CAS-Nummer: 20734-76-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...