3,5-Dimethyl-1H-pyrrol-2-carbaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR019785-1G
OR019785-5G
OR019785-10G
APOSOR019785-1GEA
21.9
EUR
APOSOR019785-1G
APOSOR019785-5G
APOSOR019785-10G
3,5-Dimethyl-1H-pyrrol-2-carbaldehyd 98%
3,5-Dimethyl-1H-pyrrole-2-carbaldehyde
Formel:
C7H9NO Schmelzpunkt: 95…99 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00111522 CAS-Nummer: 2199-58-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...