Ethyl-3,5-dichloroimidazo[1,2-a]pyridine-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR01979-1G OR01979-250MG
APOSOR01979-1GEA 346 EUR
APOSOR01979-1G APOSOR01979-250MG
Ethyl-3,5-dichloroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl-3,5-dichloroimidazo[1,2-a]pyridine-2-carboxylate
Formel: C₁₀H₈Cl₂N₂O₂
Molecular Weight: 259.09 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09842623
CAS-Nummer: 1000018-01-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR