Ethyl-3-bromo-5-chloroimidazo[1,2-a]pyridine-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR01981-1G
OR01981-250MG
APOSOR01981-1GEA
474
EUR
APOSOR01981-1G
APOSOR01981-250MG
Ethyl-3-bromo-5-chloroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl-3-bromo-5-chloroimidazo[1,2-a]pyridine-2-carboxylate
Formel:
C₁₀H₈BrClN₂O₂ Molecular Weight: 303,55 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864799 CAS-Nummer: 1000018-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...