3-Amino-4-iod-1H-indazol

Lieferant: Apollo Scientific

Synonyme: 4-Iodo-1H-indazol-3-amine

OR01986-1G OR01986-5G
APOSOR01986-1GEA 58 EUR
APOSOR01986-1G APOSOR01986-5G
3-Amino-4-iod-1H-indazol
3-Amino-4-iodo-1H-indazole
Formel: C₇H₆IN₃
Molecular Weight: 259,0471 g/mol
Schmelzpunkt: 150…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03426697
CAS-Nummer: 599191-73-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR