1-Methyl-4-phenoxy-1H-indazol-3-amine
Lieferant: Apollo Scientific
Synonyme:
3-Amino-1-methyl-4-phenoxy-1H-indazole
OR01989-1G
OR01989-250MG
APOSOR01989-1GEA
219
EUR
APOSOR01989-1G
APOSOR01989-250MG
1-Methyl-4-phenoxy-1H-indazol-3-amine
1-Methyl-4-phenoxy-1H-indazol-3-amine
Formel:
C₁₄H₁₃N₃O Molecular Weight: 239,28 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864805 CAS-Nummer: 1000018-07-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...