6-Chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine5-carbaldehyde

Lieferant: Apollo Scientific
Achtung

OR01992-100MG OR01992-500MG OR01992-1G
APOSOR01992-100MGEA 29 EUR
APOSOR01992-100MG APOSOR01992-500MG APOSOR01992-1G
6-Chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine5-carbaldehyde
6-Chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine5-carbaldehyde
Formel: C₉H₈ClN₃O
Molecular Weight: 209,64 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08691397
CAS-Nummer: 898911-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR