4'-(Bromacetyl)acetanilid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Acetamidophenacyl bromide

OR01996-1G OR01996-5G OR01996-10G
APOSOR01996-1GEA 92.8 EUR
APOSOR01996-1G APOSOR01996-5G APOSOR01996-10G
4'-(Bromacetyl)acetanilid
4'-(Bromacetyl)acetanilid
Formel: C₁₀H₁₀BrNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00728749
CAS-Nummer: 21675-02-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR