tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate, 5-BOC-2-AMINO-4,5,6,7-TETRAHYDROTHIAZOLO[5,4-C]PYRIDINE, 2-Amino-6,7-dihydro-4H-thiazolo[5,4-c]pyridine-5-carboxylicacidtert-butylester, tert-Butyl 2-amino-6,7-dihydro[1,3]thiazolo[5,4-c]pyridine-5(4H)-carboxylate

OR01999-5G
APOSOR01999-5GEA 156 EUR
APOSOR01999-5G
tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate 97%
tert-Butyl-2-amino-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate
Formel: C₁₁H₁₇N₃O₂S
Molecular Weight: 255,336 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08448157
CAS-Nummer: 365996-05-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR