4-(Methylsulphonyl) phenylacetonitrile

Lieferant: Apollo Scientific
Achtung

OR0210-1G OR0210-250MG
APOSOR0210-1GEA 79.9 EUR
APOSOR0210-1G APOSOR0210-250MG
4-(Methylsulphonyl) phenylacetonitrile
4-(Methylsulphonyl) phenylacetonitrile
Formel: C₉H₉NO₂S
Molecular Weight: 195,24 g/mol
Schmelzpunkt: 122…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00079775
CAS-Nummer: 25025-07-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR