Cbz-Tyrosinhydrat

Lieferant: Apollo Scientific

Synonyme: Z-Tyr-OH, N-Cbz-L-tyrosine, Cbz-Tyr-OH

OR0222-100G OR0222-25G
APOSOR0222-100GEA 115 EUR
APOSOR0222-100G APOSOR0222-25G
Cbz-Tyrosinhydrat
Cbz-Tyrosinhydrat
Formel: C₁₇H₁₇NO₅
Molecular Weight: 315.33 g/mol
Schmelzpunkt: 93…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00037180
CAS-Nummer: 1164-16-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR