1-(3-Cyanbenzyl)piperazin 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-[(Piperazin-1-yl)methyl]benzonitrile

OR0229-1G OR0229-5G
APOSOR0229-1GEA 114 EUR
APOSOR0229-1G APOSOR0229-5G
1-(3-Cyanbenzyl)piperazin 97%
1-(3-Cyanbenzyl)piperazin
Formel: C₁₂H₁₅N₃
Molecular Weight: 201,27 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 203047-38-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR