1-(cyclopropylcarbonyl)piperazine hydrochloride 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Cyclopropyl(piperazin-1-yl)methanone hydrochloride, 1-(Cyclopropylcarbonyl)piperazine hydrochloride

OR0246-5G OR0246-25G OR0246-100G
APOSOR0246-5GEA 51 EUR
APOSOR0246-5G APOSOR0246-25G APOSOR0246-100G
1-(cyclopropylcarbonyl)piperazine hydrochloride 97%
1-(cyclopropylcarbonyl)piperazine hydrochloride
Formel: C₈H₁₅ClN₂O
Molecular Weight: 190.671 g/mol
Schmelzpunkt: 174…176 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03428576
CAS-Nummer: 1021298-67-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR