3-Chlor-4-methylbenzylamin
Lieferant: Apollo Scientific
|
Gefahr
|
OR0253-1G
OR0253-5G
APOSOR0253-1GEA
0
EUR
APOSOR0253-1G
APOSOR0253-5G
3-Chlor-4-methylbenzylamin
3-Chlor-4-methylbenzylamin
Formel:
C₈H₁₀ClN Siedepunkt: 128…130 °C/15mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014816 CAS-Nummer: 67952-93-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...