6-Chlor-4-nitro-o-toluidin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chlor-6-methyl-4-nitroanilin, 2-Amino-3-chlor-5-nitrotoluol, 6-Chlor-2-methyl-4-nitroanilin
OR0258-1G
OR0258-25G
OR0258-5G
APOSOR0258-1GEA
21.9
EUR
APOSOR0258-1G
APOSOR0258-25G
APOSOR0258-5G
6-Chlor-4-nitro-o-toluidin
6-Chlor-4-nitro-o-toluidin
Formel:
C₇H₇ClN₂O₂ Schmelzpunkt: 170…176 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01313703 CAS-Nummer: 69951-02-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...