3,4-Diaminobenzonitril 95%

Lieferant: Apollo Scientific
Achtung

OR0330-1G OR0330-25G OR0330-5G
APOSOR0330-1GEA 21.9 EUR
APOSOR0330-1G APOSOR0330-25G APOSOR0330-5G
3,4-Diaminobenzonitril 95%
3,4-Diaminobenzonitril
Formel: C₇H₇N₃
Molecular Weight: 133,15 g/mol
Schmelzpunkt: 144…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00723901
CAS-Nummer: 17626-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR