2,7-Dichlorfluoren

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,7-Dichloro-9H-fluorene

OR0350-25G OR0350-5G OR0350-100G
APOSOR0350-25GEA 24.8 EUR
APOSOR0350-25G APOSOR0350-5G APOSOR0350-100G
2,7-Dichlorfluoren
2,7-Dichlorfluoren
Formel: C₁₃H₈Cl₂
Molecular Weight: 235,11 g/mol
Schmelzpunkt: 126…128 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00032840
CAS-Nummer: 7012-16-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR