3,5-Dichlorsalicylaldehyd
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Dichloro-2-hydroxybenzaldehyde
OR0351-100G
APOSOR0351-100GEA
94.4
EUR
APOSOR0351-100G
3,5-Dichlorsalicylaldehyd
3,5-Dichlorsalicylaldehyd
Formel:
C₇H₄Cl₂O₂ Molecular Weight: 191,01 g/mol Schmelzpunkt: 95…97 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003320 CAS-Nummer: 90-60-8 EINECS: 202-005-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...