2-Amino-6-chloropurine riboside

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Chloroguanineriboside, (2R,3R,4S,5R)-2-(2-amino-6-chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 2-Amino-6-chloro-9-(beta-D-ribofuranosyl)purine

OR0351T-1G OR0351T-5G
APOSOR0351T-1GEA 49.3 EUR
APOSOR0351T-1G APOSOR0351T-5G
2-Amino-6-chloropurine riboside
2-Amino-6-chloropurine riboside
Formel: C₁₀H₁₂ClN₅O₄
Molecular Weight: 301.69 g/mol
Schmelzpunkt: 166…168 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00005735
CAS-Nummer: 2004-07-1
EINECS: 217-905-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR