2',4'-Dichlor-6'-methylacetanilid
Lieferant: Apollo Scientific
OR0352-1G
OR0352-5G
OR0352-10G
APOSOR0352-1GEA
105
EUR
APOSOR0352-1G
APOSOR0352-5G
APOSOR0352-10G
2',4'-Dichlor-6'-methylacetanilid
2',4'-Dichlor-6'-methylacetanilid
Formel:
C₉H₉Cl₂NO Molecular Weight: 218,08 g/mol Schmelzpunkt: 174…176 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
61655-97-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...