1-Aminocyclopropancarbonsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Amino-1-cyclopropanecarboxylic acid
OR0355T-5G
OR0355T-100G
OR0355T-25G
APOSOR0355T-5GEA
33.5
EUR
APOSOR0355T-5G
APOSOR0355T-100G
APOSOR0355T-25G
1-Aminocyclopropancarbonsäure 97%
1-Aminocyclopropanecarboxylic acid
An NMDA receptor agonist acting at the glycine site.
Formel:
C₄H₇NO₂ Molecular Weight: 101,11 g/mol Schmelzpunkt: 229…231 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00009944 CAS-Nummer: 22059-21-8 EINECS: 606-917-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...