Methyl-1-aminocyclopropancarboxylathydrochlorid 97%
Lieferant: Apollo Scientific
Achtung
Synonyme: 1-Aminocyclopropane-1-carboxylic acid Methyl Ester Hydrochloride, 1-Aminocyclopropanecarboxylic acid methyll ester hydrochloride, Methyl 1-aminocyclopropanecarboxylate hydrochloride, 1-Aminocyclopropanecarboxylic acid methyl ester hydrochloride, Methyl 1-aminocyclopropanecarboxylate HCl
OR0360T-25G
APOSOR0360T-25GEA 63.8 EUR
APOSOR0360T-25G
Methyl-1-aminocyclopropancarboxylathydrochlorid 97%
Methyl-1-aminocyclopropanecarboxylate hydrochloride
Formel: C₅H₁₀ClNO₂
Molecular Weight: 151,5914 g/mol
Schmelzpunkt: 185…186 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00078879
CAS-Nummer: 72784-42-0