2,6-Dichlorisonicotinsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Dichloropyridine-4-carboxylic acid

OR0368-100G OR0368-25G OR0368-5G
APOSOR0368-100GEA 212 EUR
APOSOR0368-100G APOSOR0368-25G APOSOR0368-5G
2,6-Dichlorisonicotinsäure 98%
2,6-Dichlorisonicotinsäure
Formel: C₆H₃Cl₂NO₂
Molecular Weight: 192 g/mol
Schmelzpunkt: 208…212 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00234147
CAS-Nummer: 5398-44-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR