Diethyl(2,4-dichlorphenyl)malonat

Lieferant: Apollo Scientific
Achtung

OR0372-1G OR0372-5G OR0372-10G
APOSOR0372-1GEA 174 EUR
APOSOR0372-1G APOSOR0372-5G APOSOR0372-10G
Diethyl(2,4-dichlorphenyl)malonat
Diethyl(2,4-dichlorphenyl)malonat
Formel: C₁₃H₁₄Cl₂O₄
Molecular Weight: 305,16 g/mol
Siedepunkt: 136…139 °C/0,05mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00831103
CAS-Nummer: 111544-93-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR